MilliporeSigma™ Human Adipose Mesenchymal Stem Cell Kit
provides both cryopreserved human adipose mesenchymal stem cells (MSCs) and low-serum expansion media that is recommended for optimal growth and expansion of adipose MSCs.
Supplier:  MilliporeSigma™ SCR038
Catalog No. SCR038
Explore available promotions
Description
Description
Specifications
Specifications
Store in liquid nitrogen
Cell culture, cell differentiation, stem cell culture
1 x 106 and 500mL optimized expansion medium/Kit