MilliporeSigma™ Immobilon™-PSQ PVDF Membranes
For Western blots and protein sequencing
Supplier: MilliporeSigma™ ISEQ09120
Description
Immobilon-PSQ Membrane:Specifications
9 cm | |
For sequencing low-abundance or low-molecular weight (<20,000 Daltons) proteins | |
Immobilon-PSQ | |
9 x 12 cm |
12 cm | |
Sheet | |
0.2 μm |