missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MilliporeSigma™ Immobilon™-PSQ PVDF Membranes

For Western blots and protein sequencing

Supplier:  MilliporeSigma™ ISEQ07850

 View more versions of this product

Catalog No. ISEQ07850


Explore available promotions

Description

Description

Immobilon-PSQ Membrane:
  • 0.2 μm pore size and a large internal structure
  • Higher protein adsorption and sequencing yields than other membranes
  • Recommended for blotting proteins less than 20 kDa
  • Prevents blow-through of low molecular weight proteins
  • Specifications

    Specifications

    7 cm
    For sequencing low-abundance or low-molecular weight (<20,000 Daltons) proteins
    ISEQ
    3.30 in.
    Immobilon-PSQ
    0.2 μm
    8.4 cm
    White
    Plain
    Sheet
    2.75 in.
    7 x 8.4 cm
    Videos
    SDS
    Documents

    Documents

    Promotions

    Promotions