MilliporeSigma™ Milli-Q™ Ultrapure Water Systems Accessory, Cartridges
Choose from a wide selection of replacement cartridges
Supplier: MilliporeSigma™ QGARD00R1
Description
Choose from a wide selection of replacement cartridges
Filter Types
- Activated carbon
- Ion exchange
- Membrane
Specifications
Elix, Reverse Osmosis, Distilled | |
Milli-Q Systems | |
Replacement Cartridge |
Ion Exchange, Activated Carbon | |
Q-Gard 1 |