MilliporeSigma™ Millicell™ Cell Culture Insert Plates
Designed to support cell growth and differentiation of endothelial and epithelial cell lines
Supplier: MilliporeSigma™ PSHT004S5
Description
- For easier pipetting and isolateral access
- Can be used manually or with automated cell seeding, feeding and washing systems
- Optimized to grow and sustain high-integrity cell monolayers
- Membrane plates have feet to elevate the plate above the work surface when disassembled from the feeder tray to reduce the risk of monolayer contamination
- Tear-drop shaped receiver wells eliminate air bubbles as plates are assembled
- Isopore membrane is a polycarbonate, track-etched screen filter recommended for all analyses in which the sample is viewed on the surface of the membrane
Type and number of components vary by model:
PSHT004R1 Includes
- One 96-Well Cell Culture Plate
- One Feeder Tray
- One 96-Well Receiver Tray
- One Lid
- Five 96-Well Culture Plates
- Five Single-Well Feeder Trays
- Five Lids
- Five 96-Well Cell Culture Plates
- Ten 96-Well Receiver Trays
- Five Lids
- Cell Attachment
- Cell Culture
- Cell Differentiation
- Drug Transport
- Permeability
- Cell-Based Assays
- Caco-2 Cell Culture
Specifications
PCF | |
Single-well feeder tray and lid | |
Sterile |
0.4 μm | |
Cell Attachment, Cell Culture, Cell Differentiation, Drug Transport | |
96 |