Learn More
MilliporeSigma™ Millivac™ Mini Pump
Compact, maintenance-free pumps that provide a consistent source of vacuum for filtration and other laboratory applications
Supplier:  MilliporeSigma™ XX5411560
 View more versions of this product
Catalog No. XX5411560
Only null left
Explore available promotions
Description
Description
Compact, maintenance-free pumps that provide a consistent source of vacuum for filtration and other laboratory applications
  • A patented diaphragm design has reduced the footprint of the pump compared to conventional models
  • Use Millivac Mini Pump to filter water and aqueous solutions
  • No lubrication required
  • Not explosion-proof
  • 4mm ID tubing
  • Dimensions: 22.6 x 9 x 14.1cm, L x W x H
Specifications
Specifications
4.1 lb.
Hose connector
Ryton (Pump Head), EPDM (Membrane), Fluoroelastomer (Valves)
1.9 kg
60 Hz
14.1 cm
22.6 cm
1.9 kg
115 V 60 Hz
4.1 lb.
6 L/min.
5.5 in.
8.8 in.
115 V
Product Suggestions
Product Suggestions