missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ MQuant™ Potassium Test Strips
High-tech, fast and simple means for semiquantitative testing
Supplier: MilliporeSigma™ 1179850001
Includes: Reagent
Description
Uses a dipicrylamine method.
Small and easy to handle for on-site use
Fast results
Ideal for screening
PET film and low reagent content for easier disposal
Graduations: 250-450--700-1000-1500mg/L
Order Info
No. of tests: 100
Specifications
| Colorimetric | |
| 1 bottle reagent K-1, 1 test vessel | |
| 250 to 1500 mg/L | |
| Dipicrylamine |
| 250-450-700-1000-1500mg/L | |
| 100 Tests | |
| 15°C to 25°C | |
| Potassium Test Strip |