MilliporeSigma™ RC800 Mini-Reservoir
Accessory for simplified stirred cell operation
Supplier:  MilliporeSigma™ 6028
Catalog No. 6028
Explore available promotions
Description
Description
  • Graduated acrylic body
  • Acetal cover, removable for easy cleaning
  • Acetal fittings, Buna-N, silicone O-rings
  • Safety valve for slow pressure bleed (max. pressure: 5 bar)
  • Polyethylene tubing
  • Wrap-around clamp for extra safety
  • Chemically sanitizable