MilliporeSigma™ SeQuant™ ZIC-HILIC HPLC Metal-free PEEK Guard Columns and Kit
PEEK coated HPLC column
Supplier: MilliporeSigma™ 1504340001
Description
SeQuant™ ZIC™-HILIC is the ideal choice for separation of polar and hydrophilic compounds. This guard column protects your analytical separation column from contamination.Specifications
1 mm | |
5 μm | |
200 AÌ | |
Fitting Guard Column |
14 mm | |
200 Å | |
5/Pk. |