MilliporeSigma™ Upstate MAP Kinase/Erk Assay Kit
Non-radioactive kit to measure phosphotransferase activity of MAP (mitogen-activated protein) Kinase in immunoprecipitates and column fractions
Supplier: MilliporeSigma™ 17191
Includes: Components of Kit
- Anti-phospho-MBP, clone P12
- Assay Dilution Buffer I (ADBI)
- MAP Kinase/Erk Substrate Cocktail II
- Inhibitor Cocktail
- Magnesium/ATP Cocktail
Specifications
Kinase Assay | |
Anti-phospho-MBP, clone P12, Assay Dilution Buffer I (ADBI), MAP Kinase/Erk Substrate Cocktail II, Inhibitor Cocktail, Magnesium/ATP Cocktail | |
1 Kit | |
Kinase Assay Kit |