missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MilliporeSigma™ EMD Human Epidermal Keratinocytes, Neonatal
provide an ideal serum-free culture model for the study of wound healing, toxicology, or biology when grown in our recommended medium.
Supplier: MilliporeSigma™ SCCE020
Description
- Millipore’s Human Neonatal Epidermal Keratinocytes, when grown in our recommended medium, provide an ideal serum-free culture model for the study of wound healing, toxicology, or biology
- These cells are not exposed to antimicrobials or phenol red when cultured in EpiGRO™ Human Keratinocyte Complete Media Kit
- Cryopreserved at passage 1 to ensure highest viability and plating efficiency
- When used properly this product should scale for a minimum of 15 population doublings
Specifications
-150°C | |
Cell Culture | |
Human Neonatal Keratinocyte |
Cells are provided cryopreserved in 1mL of 10% FBS with 10% DMSO | |
500,000 cells (5 x 105 cells/vial) |
For research use only. Not for use in diagnostic procedures.