missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Emtricitabine 98.0+%, TCI America™
$95.16 - $532.87
Chemical Identifiers
CAS | 143491-57-0 |
---|---|
Molecular Formula | C8H10FN3O3S |
Molecular Weight (g/mol) | 247.244 |
MDL Number | MFCD00870151 |
InChI Key | XQSPYNMVSIKCOC-NTSWFWBYSA-N |
PubChem CID | 60877 |
ChEBI | CHEBI:31536 |
IUPAC Name | 4-amino-5-fluoro-1-[(2R,5S)-2-(hydroxymethyl)-1,3-oxathiolan-5-yl]pyrimidin-2-one |
SMILES | C1C(OC(S1)CO)N2C=C(C(=NC2=O)N)F |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
E100725MG
|
TCI America
E100725MG |
25 mg |
Each for $95.16
|
|
|||||
E1007250MG
|
TCI America
E1007250MG |
250 mg |
Each for $532.87
|
|
|||||
Chemical Identifiers
143491-57-0 | |
247.244 | |
XQSPYNMVSIKCOC-NTSWFWBYSA-N | |
CHEBI:31536 | |
C1C(OC(S1)CO)N2C=C(C(=NC2=O)N)F |
C8H10FN3O3S | |
MFCD00870151 | |
60877 | |
4-amino-5-fluoro-1-[(2R,5S)-2-(hydroxymethyl)-1,3-oxathiolan-5-yl]pyrimidin-2-one |
Specifications
143491-57-0 | |
White-Yellow | |
MFCD00870151 | |
XQSPYNMVSIKCOC-NTSWFWBYSA-N | |
4-amino-5-fluoro-1-[(2R,5S)-2-(hydroxymethyl)-1,3-oxathiolan-5-yl]pyrimidin-2-one | |
60877 | |
247.24 | |
Crystalline Powder |
155°C | |
C8H10FN3O3S | |
25 mg | |
C1C(OC(S1)CO)N2C=C(C(=NC2=O)N)F | |
247.244 | |
CHEBI:31536 | |
≥98.0% (HPLC,T) | |
Emtricitabine |
Safety and Handling
RTECSNumber : UW7360500
TSCA : No