missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Enalaprilat, Thermo Scientific Chemicals

The active metabolite of enalapril

Supplier:  Thermo Scientific Chemicals J63392MA

 View more versions of this product

Catalog No. AAJ63392MA


Explore available promotions

Description

Description

Enalapril is a ACE (angiotensin-converting enzyme) inhibitor, suggested to be more potent than Captopril (sc-200566). Studies indicate that Enalapril downregulates the expression of IL-β, IL-6, and NF-κB. Further studies show that Enalapril upregulates the expression of VEGF and heme oxygenase-1. Enalaprilat reports demonstrate that the compound has some antioxidant activity, which does not include the PKC-NADPH oxidase pathway.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Enalapril is a ACE (angiotensin-converting enzyme) inhibitor, suggested to be more potent than Captopril (sc-200566). Studies indicate that Enalapril downregulates the expression of IL-β, IL-6, and NF-κB. Further studies show that Enalapril upregulates the expression of VEGF and heme oxygenase-1. Enalaprilat reports demonstrate that the compound has some antioxidant activity, which does not include the PKC-NADPH oxidase pathway.

Solubility
Soluble in water with heating (∼60°C)

Notes
Store in a cool dry place. Keep away from oxidizing agents.
Specifications

Specifications

Enalaprilat
148°C to 151°C
C18H24N2O5
10 mg
enalaprilat, enalapril acid, enalaprilate, enalaprilatum, enalapril diacid, enalaprilic acid, enalaprilat anhydrous, n-1s-1-carboxy-3-phenylpropyl-l-alanyl-l-proline, unii-q508q118jm
LZFZMUMEGBBDTC-QEJZJMRPSA-N
(2S)-1-[(2S)-2-[[(1S)-1-carboxy-3-phenylpropyl]amino]propanoyl]pyrrolidine-2-carboxylic acid
5462501
384.42
76420-72-9
White
MFCD00865786
14,3568
Soluble in water with heating (∽60°C)
CC(C(=O)N1CCCC1C(=O)O)NC(CCC2=CC=CC=C2)C(=O)O
348.399
CHEBI:4786
Crystalline Solid

Chemical Identifiers

76420-72-9
348.399
LZFZMUMEGBBDTC-QEJZJMRPSA-N
5462501
(2S)-1-[(2S)-2-[[(1S)-1-carboxy-3-phenylpropyl]amino]propanoyl]pyrrolidine-2-carboxylic acid
C18H24N2O5
MFCD00865786
enalaprilat, enalapril acid, enalaprilate, enalaprilatum, enalapril diacid, enalaprilic acid, enalaprilat anhydrous, n-1s-1-carboxy-3-phenylpropyl-l-alanyl-l-proline, unii-q508q118jm
CHEBI:4786
CC(C(=O)N1CCCC1C(=O)O)NC(CCC2=CC=CC=C2)C(=O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 278-459-3

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only