Encon™ Hydrosep Water Treatment Additive for Portable Eyewash Stations
Supplier:  Encon™ 01110764
Catalog No. 19-021780
Explore available promotions
Description
Description
Specifically formulated to provide protection against bacteria, fungi, algae, staph, and acanthamoeba in self-contained eyewash stations. One 8 oz. (237mL) bottle of Hydrosep will protect 5 to 20 gal. (18.9 to 75.7L) of potable water for up to 120 days.
Order Info
Supplied four 8 oz. (237mL) bottles to a case.