AirClean™ Systems Endeavour™ Ductless Fume Hood, Sliding Sash
For increased user protection, the AirClean Systems Endeavour™ Ductless Fume Hood with Sliding Sash features thermally fused polypropylene construction that promotes chemical resistance and spill containment.
$29442.86 - $29442.86
Specifications
Product Type Ductless Fume Hood
View More Specs
Alert:
Requires an air filter in order to operate. Air filter must be purchased separately.
Products 1
Catalog Number Mfr. No. Dimensions (D x W x H) Price Quantity  
Catalog Number Mfr. No. Dimensions (D x W x H) Price Quantity  
36-100-0070
View Documents
AirClean™ Systems
ACPT6000S
25 x 66 x 36 in. (63.5 x 167.6 x 91.4 cm) N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
  • Large capacity filtration bed to handle a wide variety of applications
  • AirZone baffling promotes uniform airflow and containment
  • Microprocessor controller with variable airflow controller and constant filter monitoring
  • Integral vapor proof fluorescent lighting for effective work area illumination
  • Built-in solid state gas detection
  • Shipped fully assembled, filters installed
  • Optional fixtures and services can be easily placed on front fascia of hood
Available Options:
  • 110 V or 220 V models
  • Polypropylene base cabinets, vented and unvented models
  • Slip hatch electrical cord access ports
  • Polypropylene sinks and deck mounted fixtures
  • Sturdy cart/trolley with locking casters for mobility
  • Epoxy coated metal stand with adjustable leveling feet
Certifications
UL listed, CSA, CE marked
Compliance
Meets all relevant OSHA, ANSI Z9.5 and ASHRAE 110 standards
Specifications
Specifications
Ductless Fume Hood
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'