missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

endo-N-Hydroxy-5-norbornene-2,3-dicarboximide, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1320530

 View more versions of this product

Catalog No. AAA1320530



Description

Description

It decreases racemization in peptide synthesis and inhibits formation of N-acylureas.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It decreases racemization in peptide synthesis and inhibits formation of N-acylureas.

Solubility
Soluble in ethanol

Notes
Store at room temperature. Incompatible with oxidizing agents.
Specifications

Specifications

endo-N-Hydroxy-5-norbornene-2,3-dicarboximide
166°C to 170°C
MFCD00065691
13540
Soluble in ethanol
ON1C(=O)C2C3CC(C=C3)C2C1=O
179.18
179.18
21715-90-2
C9H9NO3
250 g
honb, n-hydroxy-5-norbornene-2,3-dicarboximide, 2-hydroxy-3a,4,7,7a-tetrahydro-1h-4,7-methanoisoindole-1,3 2h-dione, 4-hydroxy-4-azatricyclo 5.2.1.0 2,6 dec-8-ene-3,5-dione, n-hydroxynorborn-5-ene-2,3-dicarboximide, n-hydroxy-5-norbornene-2,3-dicarboxylic imide, 5-norbornene-2, n-hydroxy, 4,7-methano-1h-isoindole-1,3 2h-dione, 3a,4,7,7a-tetrahydro-2-hydroxy, 4,3 2h-dione, 3a,4,7,7a-tetrahydro-2-hydroxy, 4-hydroxy-4-azatricyclo 5.2.1.0<2,6> dec-8-ene-3,5-dione
ZUSSTQCWRDLYJA-UHFFFAOYNA-N
4-hydroxy-4-azatricyclo[5.2.1.0²,⁶]dec-8-ene-3,5-dione
89529
97%

Chemical Identifiers

21715-90-2
179.18
ZUSSTQCWRDLYJA-UHFFFAOYNA-N
89529
ON1C(=O)C2C3CC(C=C3)C2C1=O
C9H9NO3
MFCD00065691
honb, n-hydroxy-5-norbornene-2,3-dicarboximide, 2-hydroxy-3a,4,7,7a-tetrahydro-1h-4,7-methanoisoindole-1,3 2h-dione, 4-hydroxy-4-azatricyclo 5.2.1.0 2,6 dec-8-ene-3,5-dione, n-hydroxynorborn-5-ene-2,3-dicarboximide, n-hydroxy-5-norbornene-2,3-dicarboxylic imide, 5-norbornene-2, n-hydroxy, 4,7-methano-1h-isoindole-1,3 2h-dione, 3a,4,7,7a-tetrahydro-2-hydroxy, 4,3 2h-dione, 3a,4,7,7a-tetrahydro-2-hydroxy, 4-hydroxy-4-azatricyclo 5.2.1.0<2,6> dec-8-ene-3,5-dione
4-hydroxy-4-azatricyclo[5.2.1.0²,⁶]dec-8-ene-3,5-dione
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 244-538-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only