FUJIFILM Wako Endotoxin Specific Buffer
Endotoxin-specific buffer functions as a beta glucan blocker when used with untreated LAL reagents
Supplier:  FUJIFILM Wako ESB0006
Catalog No. 12-333-054
Explore available promotions
Description
Description
  • Each vial has solution for use in reconstituting a max 5.2 mL of reagent
  • Ideal for LAL that has the potential to react with glucans
Specifications
Specifications
Lyophilized
LAL reagents
6 Vials
6 mL
6 x 6 mL vials