Learn More
Superior Glove™ Endura™ Oilbloc™ Arc-Flash Goat-Grain Leather Drivers Gloves
Goatskin leather provides abrasion resistance, while Oilbloc™ treatment ensures oil and water resistance.
$31.10 - $31.10
Specifications
Arc Flash Level 4
Color Brown
Cut Resistance Rating ANSI A4
Hand Specific
Material Grain Goatskin Leather
View More Specs
Products 1
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
17-401-278
View Documents
Superior Glove™
378GOBKLL
Large
Pair of 12 for $31.10
Case of 60 Pair for $1,781.40
Only null left
Description
Description
  • Goatskin leather has a high tensile strength for increased abrasion resistance 
  • Treated with Oilbloc™ for oil and water resistance
  • Aramid Fiber provides 360° cut protection 
  • Drivers style design slips on easily with an elastic back for a snug fit 
  • Arc flash tested for level 4 with an ATPV of 41 cal/cm2
Specifications
Specifications
Level 4
ANSI A4
Grain Goatskin Leather
Automotive Assembly, Construction, Manufacturing, and Utility Work
Aramid Fiber
378GOBKL
Brown
Specific
Driver Gloves
Elasticated
ANSI/ISEA 105-16 Level 4
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'