missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
ENG Scientific Lactophenol Cotton Blue Stain, Certified
For staining fungi using Lactophenol Cotton Blue Stain.
$873.32 - $873.32
Specifications
Type | Lactophenol Cotton Blue Stain |
---|---|
Includes | Phenol, lactic acid, glycerol, aniline blue |
Format | Bottle |
For Use With (Application) | Staining fungi, in-vitro diagnostic use |
Stain Type | Lactophenol Cotton Blue |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
ES6730
|
ENG Scientific
ES6730 |
2 x 100 mL |
Pack of 2 for $873.32
|
|
|||||
Description
- Lactic acid acts as a preservative for fungi, killed by phenol and stained by cotton blue
- Fungal elements are stained a deep blue; background is pale blue
- Lactophenol Cotton Blue Stain
- Includes Phenol, lactic acid, glycerol, aniline blue
- For use with staining fungi, in-vitro diagnostic use/li>
Specifications
Lactophenol Cotton Blue Stain | |
Bottle | |
Lactophenol Cotton Blue |
Phenol, lactic acid, glycerol, aniline blue | |
Staining fungi, in-vitro diagnostic use |
For in vitro diagnostic use only.