Learn More
English/Metric Meter Sticks
Available with plain or metal ends.
$15.26 - $25.33
Specifications
Products 2
Catalog Number Mfr. No. Price Quantity  
Catalog Number Mfr. No. Price Quantity  
S40625
View Documents
Customer Channels Group (Ccg)
S40625
N/A
Only null left
S40626
View Documents
Customer Channels Group (Ccg)
R690B
N/A
Only null left
Description
Description
  • Front is ruled in metric units with millimeter increments on both edges
  • Back is ruled in inches with 0.125 in. increments
  • Made of pine with a multiple-coat clear lacquer finish for increased durability
  • Dimensions: 1W × 0.25 in. thick (2.54 × 0.63cm)
Specifications
Specifications
0.0
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'