missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Entecavir Monohydrate 98.0+%, TCI America™
$119.30 - $359.76
Chemical Identifiers
| CAS | 209216-23-9 |
|---|---|
| Molecular Formula | C12H17N5O4 |
| Molecular Weight (g/mol) | 295.299 |
| MDL Number | MFCD09754448 |
| InChI Key | YXPVEXCTPGULBZ-WQYNNSOESA-N |
| Synonym | entecavir hydrate, entecavir monohydrate, baraclude tn, entecavir usan, entecavir hydrate jan, baraclude, 9-1s,3r,4s-4-hydroxy-3-hydroxymethyl-2-methylenecyclopentyl guanine monohydrate, entecavir inn, 2-amino-9-1s,3r,4s-4-hydroxy-3-hydroxymethyl-2-methylidenecyclopentyl-1,9-dihydro-6h-purin-6-one-water 1/1 |
| PubChem CID | 16052026 |
| ChEBI | CHEBI:59902 |
| IUPAC Name | 2-amino-9-[(1S,3R,4S)-4-hydroxy-3-(hydroxymethyl)-2-methylidenecyclopentyl]-3H-purin-6-one;hydrate |
| SMILES | C=C1C(CC(C1CO)O)N2C=NC3=C2NC(=NC3=O)N.O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
E089950MG
|
TCI America
E089950MG |
50 mg |
Each for $119.30
|
|
|||||
|
E0899200MG
|
TCI America
E0899200MG |
200 mg |
Each for $359.76
|
|
|||||
Chemical Identifiers
| 209216-23-9 | |
| 295.299 | |
| YXPVEXCTPGULBZ-WQYNNSOESA-N | |
| 16052026 | |
| 2-amino-9-[(1S,3R,4S)-4-hydroxy-3-(hydroxymethyl)-2-methylidenecyclopentyl]-3H-purin-6-one;hydrate |
| C12H17N5O4 | |
| MFCD09754448 | |
| entecavir hydrate, entecavir monohydrate, baraclude tn, entecavir usan, entecavir hydrate jan, baraclude, 9-1s,3r,4s-4-hydroxy-3-hydroxymethyl-2-methylenecyclopentyl guanine monohydrate, entecavir inn, 2-amino-9-1s,3r,4s-4-hydroxy-3-hydroxymethyl-2-methylidenecyclopentyl-1,9-dihydro-6h-purin-6-one-water 1/1 | |
| CHEBI:59902 | |
| C=C1C(CC(C1CO)O)N2C=NC3=C2NC(=NC3=O)N.O |
Specifications
| 209216-23-9 | |
| White | |
| MFCD09754448 | |
| entecavir hydrate, entecavir monohydrate, baraclude tn, entecavir usan, entecavir hydrate jan, baraclude, 9-1s,3r,4s-4-hydroxy-3-hydroxymethyl-2-methylenecyclopentyl guanine monohydrate, entecavir inn, 2-amino-9-1s,3r,4s-4-hydroxy-3-hydroxymethyl-2-methylidenecyclopentyl-1,9-dihydro-6h-purin-6-one-water 1/1 | |
| C=C1C(CC(C1CO)O)N2C=NC3=C2NC(=NC3=O)N.O | |
| 295.299 | |
| CHEBI:59902 | |
| ≥98.0% (HPLC,T) | |
| Entecavir Monohydrate |
| 259°C | |
| C12H17N5O4 | |
| 50 mg | |
| YXPVEXCTPGULBZ-WQYNNSOESA-N | |
| 2-amino-9-[(1S,3R,4S)-4-hydroxy-3-(hydroxymethyl)-2-methylidenecyclopentyl]-3H-purin-6-one;hydrate | |
| 16052026 | |
| 277.28 | |
| Crystalline Powder |
Safety and Handling
TSCA : No