Sartorius™ Entris™ II Essential Analytical Balance
Entris™ II Essential line of balances offers highly accurate results via monolithic weigh cell technology, fastest stabilization time in its class using state-of -the-art weighing sensors and high chemical resistance.
$2390.50 - $3349.90
Specifications
Certifications/Compliance CSA number 80000336/CE certificate
Connection Type 9 Pin USB C
Display Type Intuitive, wear resistant LED screen with touch technology
Readability 0.1 mg
Includes Weighing pan, Pan support, Power supply unit with country-specific adapters, Draft shield, For models with analytical draft shield
View More Specs
Products 6
Catalog Number Mfr. No. Capacity Calibration Price Quantity  
Catalog Number Mfr. No. Capacity Calibration Price Quantity  
14558810
View Documents Promotion Details
Sartorius™
BCE641S
60 g No internal calibration
Each for $2,390.50
14558811
View Documents Promotion Details
Sartorius™
BCE64I1S
60 g Internal
Each for $2,630.35
14558781
View Documents Promotion Details
Sartorius™
BCE1241S
120 g No internal calibration
Each for $2,774.27
14558782
View Documents Promotion Details
Sartorius™
BCE124I1S
120 g Internal
Each for $2,990.13
14558790
View Documents Promotion Details
Sartorius™
BCE2241S
220 g No internal calibration
Each for $3,134.03
14558791
Accessories View Documents Promotion Details
Sartorius™
BCE224I1S
220 g Internal
Each for $3,349.90
Description
Description
Entris™ II Essential line of balances offers highly accurate results via monolithic weigh cell technology, fastest stabilization time in its class using state-of -the-art weighing sensors and high chemical resistance.
  • High quality standards engineered in Germany
  • Excellent repeatability with rectangular weighing pan
  • Overload protection
  • Rugged design weighs up to a pre-set amount
  • High chemical resistance
  • Crafted from hard-wearing polybutylene terephthalate (PBT), stainless steel and glass
  • Prevents cross contamination
  • Wipe-clean design
  • Easy-to-remove parts
  • Effective draft shield
  • Minimizes weighing errors caused by electrostatically charged samples using specially coated glass parts while isoCAL (internal calibration and adjustment) for total assurance of accurate weighing results
  • Assures (SOP) compliant operation with self-notification if calibration is outside the normal range
  • Combines intuitive, wear resistant LED and touch technology
  • Plug-and-play convenience
  • Data output for dynamic weighing applications
  • Easily adapts to your ambient conditions
  • 12 built-in applications
  • GLP / GMP compliant printout
  • Modern connectivity methods
  • Password protection for secure operation
Specifications
Specifications
CSA number 80000336/CE certificate
Intuitive, wear resistant LED screen with touch technology
Weighing pan, Pan support, Power supply unit with country-specific adapters, Draft shield, For models with analytical draft shield
Analytical Balances
1.5 sec.
317 mm
345 mm
9 Pin USB C
0.1 mg
PBT, Glass, Stainless Steel
0.1 mg
Analytical Balance
219 mm
100/240 VAC (±10%)
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'