Covidien EnviroStar™ Noninfectious Waste Sharps Disposal Containers
Manufactured with recyled materials, these containers are designed to reduce injuries and disposal costs for non-contaminated sharps
Supplier: Covidien 8781
Description
- Designed to contain sharps that can cause a percutaneous tear but are not infected with bloodborne pathogens, such as light bulbs, broken slides or Petri dishes, and glass lab bottles
- Environmental green container color helps to differentiate from other disposal containers and promotes proper waste segregation
- Made with environmentally friendly recycled material, enhancing Reduce-Recycle-Reuse programs
Specifications
30.3 L | |
Green | |
Autoclavable |
8 gal. | |
EnviroStar | |
For sharps not infected with blood-borne pathogens |