missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific™ Enzyme Freezer Storage Bins
Store and manage inventory effectively with Enzyme Freezer Storage Bins.
Supplier: Thermo Scientific™ 6736
Description
Store and manage inventory effectively with Thermo Scientific™ Enzyme Freezer Storage Bins.
- High-impact ABS plastic, off-white
- Interior: 3.5W × 3D × 24in.L (90 × 76 ×610mm)
- Exterior: 4.4W × 3.04 × 25.25in.L (112 × 77 × 641mm)
- Labels not included
Specifications
ABS Plastic Enzyme Freezer Storage Bin | |
3.04 in. | |
25.25 in. | |
112 in. |
Storage and inventory management | |
77 mm | |
641 mm | |
4.4 mm |