Learn More
Eosin B, Thermo Scientific Chemicals
$85.10 - $224.62
Chemical Identifiers
CAS | 548-24-3 |
---|---|
Molecular Formula | C20H6Br2N2Na2O9 |
MDL Number | MFCD00005041 |
Synonym | C.I. 45400; 4,5-Dibromo-2,7-dinitrofluorescein disodium salt |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1737714
|
Thermo Scientific Chemicals
A1737714 |
25 g |
Each for $85.10
|
|
|||||
AAA1737722
|
Thermo Scientific Chemicals
A1737722 |
100 g |
Each for $224.62
|
|
|||||
Description
Eosin B sodium salt is a cell and tissue stain. Eosin B sodium salt is used as a counter stain for collagen, as a tissue stain for cell granules, nuclei, and microorganisms when coupled with Azure A (sc-203729), and as a stain for cell differentiation of the anterior pituitary.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEosin B sodium salt is a cell and tissue stain. Eosin B sodium salt is used as a counter stain for collagen, as a tissue stain for cell granules, nuclei, and microorganisms when coupled with Azure A (sc-203729), and as a stain for cell differentiation of the anterior pituitary.
Solubility
Soluble in water (390 g/L at 20°C), and ethanol.
Notes
Store at room temperature. Keep away from strong oxidizing agents.
Chemical Identifiers
548-24-3 | |
C20H6Br2N2Na2O9 | |
MFCD00005041 | |
C.I. 45400; 4,5-Dibromo-2,7-dinitrofluorescein disodium salt |
Specifications
548-24-3 | |
MFCD00005041 | |
C.I. 45400; 4,5-Dibromo-2,7-dinitrofluorescein disodium salt | |
624.08 |
C20H6Br2N2Na2O9 | |
25 g | |
Soluble in water (390g/L at 20°C),and ethanol. | |
Eosin B |
Safety and Handling
P280-P305+P351+P338-P310
H318
EINECSNumber : 208-943-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only