missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Eosin Y (Certified Biological Stain), Fisher Chemical™
Supplier: Fisher Chemical E51125
Specifications
Eosin Y | |
Pass Test | |
C20H12Br4Na2O8 | |
FD&C Red No. 22, Acid Red 87 | |
C1=CC=C(C(=C1)C2=C3C=C(C(=O)C(=C3OC4=C(C(=C(C=C24)Br)[O-])Br)Br)Br)C(=O)[O-].O.O.O.[Na+].[Na+] | |
745.904 | |
691.85 | |
Glass Bottle | |
Brown/Red | |
Solid |
17372-87-1 | |
88% min. | |
MFCD00005040 | |
MASXMTNVNASWNH-UHFFFAOYSA-L | |
disodium;2-(2,4,5,7-tetrabromo-3-oxido-6-oxoxanthen-9-yl)benzoate;trihydrate | |
91886399 | |
Certified Biological Stain | |
45380 | |
25 g |
Chemical Identifiers
17372-87-1 | |
745.904 | |
MASXMTNVNASWNH-UHFFFAOYSA-L | |
91886399 | |
C1=CC=C(C(=C1)C2=C3C=C(C(=O)C(=C3OC4=C(C(=C(C=C24)Br)[O-])Br)Br)Br)C(=O)[O-].O.O.O.[Na+].[Na+] |
C20H12Br4Na2O8 | |
MFCD00005040 | |
FD&C Red No. 22, Acid Red 87 | |
disodium;2-(2,4,5,7-tetrabromo-3-oxido-6-oxoxanthen-9-yl)benzoate;trihydrate |