missing translation for 'onlineSavingsMsg'
Learn More

Eosin Y Disodium Salt, Spectrum™ Chemical

$249.53 - $24658.79

Chemical Identifiers

CAS 17372-87-1
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18610363
View Documents
Spectrum Chemical Mfg Cor
EO11025GM
25 g Amber Glass Bottle
Each for $249.53
Only null left
 
18610362
View Documents
Spectrum Chemical Mfg Cor
EO110100GM
100 g Amber Glass Bottle
Each for $504.57
Only null left
 
18610361
View Documents
Spectrum Chemical Mfg Cor
EO1101KGBL
1 kg Amber Glass Bottle
Each for $1,611.65
Only null left
 
18610365
View Documents
Spectrum Chemical Mfg Cor
EO1105KGBL
5 kg Amber Glass Bottle
Each for $5,509.06
Only null left
 
18610364
View Documents
Spectrum Chemical Mfg Cor
EO11025KGBL
25 kg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

17372-87-1

Specifications

Solid
1
Red
691.85
Eosin Y Disodium Salt
17372-87-1
Amber Glass Bottle
25 g
Ungraded
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.