Learn More
Agilent BioTek Epoch 2 Microplate Spectrophotometer
The BioTek Epoch 2 microplate spectrophotometer delivers excellent performance for UV-Vis absorbance measurements. For Research Use Only. Not for use in diagnostic procedures.
$23011.88 - $27262.99
Specifications
Detector Type | Photodiode |
---|---|
Light Source | Xenon Flash Lamp |
Measurement Speed | 96 wells: 8 sec., 384 wells: 14 sec. |
Accuracy | ±2 nm |
Certifications/Compliance | CE and TUV marked. RoHS compliant. |
Catalog Number | Mfr. No. | Model | Includes | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Model | Includes | Price | Quantity | ||||
BTEPOCH2NS
|
Agilent BioTek
EPOCH2NSSN |
EPOCH2NS | No Touchscreen; No cuvette port |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
BTEPOCH2NSC
|
Agilent BioTek
EPOCH2NSCSN |
EPOCH2NSC | Cuvette Port |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
BTEPOCH2TS
|
Agilent BioTek
EPOCH2TSSN |
EPOCH2TS | Touchscreen only |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
BTEPOCH2TSC
|
Agilent BioTek
EPOCH2TSCSN |
EPOCH2TSC | Touchscreen; Cuvette port |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
The BioTek Epoch 2 microplate spectrophotometer delivers excellent performance for UV-Vis absorbance measurements. Measurements can be performed in 6- to 384-well microplates, cuvets, and in microvolume samples with the available Take3 microvolume plate. The broad wavelength range enables applications from nucleic acid and protein quantification in the low UV to microbial growth assays higher wavelengths
Epoch 2 is easy to use and the optional touch screen interface makes it simple to choose predefined protocols or to define custom programs. Endpoint, kinetic, spectral scanning and well area scanning modes, plus incubation and shaking enable wide-ranging applications.
Specifications
Photodiode | |
96 wells: 8 sec., 384 wells: 14 sec. | |
CE and TUV marked. RoHS compliant. | |
22 in. | |
200 to 999 nm, 1 nm increments |
Xenon Flash Lamp | |
±2 nm | |
Nucleic acid and protein quantification to ELISA and microbial growth assays | |
50/60 Hz |