Eppendorf™ 5430 Series Microcentrifuge Rotor Lids
For use with F-45-30-11 fixed angle rotor
Supplier: Eppendorf™ 022654063
Description
Ideal for basic applications as well as hazardous samplesSpecifications
Rotor Lid | |
Plastic | |
Eppendorf 5430 Microcentrifuge FA-45-30-11 Rotor | |
Autoclavable |