Eppendorf™ Adapters for Combitips Advanced™ Pipet Tips
Use these adapters with Eppendorf pipet tips. Eppendorf™ Adapters for Combitips Advanced™ Pipet Tips are color-coded and autoclavable.
$60.79 - $366.16
Catalog Number | Mfr. No. | For Use With (Equipment) | Color Code | Quantity | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | For Use With (Equipment) | Color Code | Quantity | Price | Quantity | |||
13-683-727
|
Eppendorf™
0030089715 |
25 mL Eppendorf™ Quality Tip | Red | 1 Piece |
Each for $60.79
|
|
|||
13-683-730
|
Eppendorf™
0030089740 |
50 mL Biopur™ Quality Tip | Light Gray | 7 Pieces |
Case of 7 Each for $366.16
|
|
|||
13-683-728
|
Eppendorf™
0030089723 |
50 mL Eppendorf™ Quality Tip | Light Gray | 1 Piece |
Each for $60.79
|
|
|||
13-683-729
|
Eppendorf™
0030089731 |
25 mL Biopur™ Quality Tip | Red | 7 Pieces |
Case of 7 Each for $366.16
|
|
|||
Description
Use these adapters with Eppendorf pipet tips. Eppendorf™ Adapters for Combitips Advanced™ Pipet Tips are color-coded and autoclavable.
- Available in 25 and 50mL
Specifications
Autoclavable |