Eppendorf™ Combitips™ Advanced Tips for Multipette™ Multi-dispensers - Eppendorf Quality Edge
Edge
Eppendorf's consumable of choice for all Eppendorf Multipette™ multi-dispensers. Available in 9 volume sizes (0.1 to 50 mL), Combitips advanced in 'Eppendorf Quality' dispense in increments as low as 100 mL and are ideally suited for a variety of standard lab applications. Also available in purity grades PCR clean, Biopur™ or Forensic DNA Grade.
Supplier:  Eppendorf™ 0030089448
 View more versions of this product
Catalog No. 13-683-704
Edge
Explore available promotions
Description
Description
Use these tips for comfortable handling while preventing damage to gloves. Eppendorf Combitips advanced™ Standard Pipetter Tips are optimized to meet the increased requirements of a modern laboratory, and offered in three different purity grades to support any application.
  • Combitips advanced are the ideal system components for all Eppendorf Multipettes
  • The 9 volume sizes (0.1 mL – 50 mL) offer a maximum range of dispensing volumes with increments as low as 100 nanoliters depending on the Combitip used
  • Elongated Combitips in volumes 2.5 mL, 5.0 mL and 10 mL make it possible to reach the bottom of the most common laboratory vessels
  • An individual color coding facilitates the quick identification of the desired Combitips
Specifications
Specifications
2.5 mL
Non-sterile
Not Autoclavable
Bulk, Bagged
100 Pk. (4 Bags x 25 Pk.)
Green
Positive Displacement System
CombiTips Advanced
Certified Quality
Non-filtered
Colorless
Eppendorf Quality (Non-sterile)
Extended Length, Positive Displacement
Pipette Tips
Single-use
Yes
Polypropylene