missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Eppendorf™ Mastercycler™ Nexus X2 Thermal Cyclers
Provide simple, flexible programming with choice of gradient or nongradient blocks
Supplier: Eppendorf™ 6336000023
Description
- Able to run two totally independent protocols at same time
- Two different blocks for smaller (32-well) and larger (64-well) assays
- Larger block features optional (GX2/GX2e) gradient function for PCR optimization
- Can be combined with Eppendorf's PCR Tubes, PCR Strips or divisible plates
- Intuitive graphic programming
- Universal block for strips, 0.1 mL, 0.2 mL and 0.5 mL PCR tubes
- Small footprint and E-mail notification
- flexlid concept allows use of all types of consumables with automatic height adjustment of lid
- Heating technology of the block: Peltier Elements, Triple Circuit Technology
- Interfaces : USB, Ethernet, CAN in/out
- Universal Aluminum thermal block
Mastercycler nexus X2/X2e
- Without gradient
- Lid temperature range: 37 to 100°C
- With gradient
- Gradient block: Over 12 columns
- Gradient range: 1 to 20°C
- Gradient temperature range: 30 to 99°C
Specifications
120 V, 50/60 Hz 700 W | |
Mastercycler Nexus GX2; With Gradient |