Eppendorf™ Rotor for Centrifuge 5430 / 5430 R
Eppendorf Rotor for Centrifuge 5430/5430 R, precision at its core. Maximize your centrifuge's potential with rotors crafted for efficiency. Tailor your experiments with various capacities and speeds, ensuring optimal sample processing.
Supplier:  Eppendorf™ 022654080
 View more versions of this product
Catalog No. 05-400-023
Explore available promotions
Description
Description
  • Precision-engineered for optimal sample separation.
  • All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121°C, 20 min.).
  • Accessories (adaptors and buckets) further customize your centrifuge setup.
  • Aerosol-tight and PTFE coated options available.
Certifications
Aerosol-tight rotors are tested and certified by the Health Protection Agency (Porton Down, UK)
Specifications
Specifications
FA-45-24-11-HS, Aerosol-tight - includes rotor lid
Rotor
30,130 x g
8.8 cm
Fixed
Aluminum
24 x 1.5/2.0 mL tubes
17,500 rpm
Centrifuge 5430 / 5430 R
Rotor Lid
Videos
SDS
Documents
Documents
Accessories