Eppendorf™ Rotor for Centrifuge 5430 / 5430 R
Eppendorf Rotor for Centrifuge 5430/5430 R, precision at its core. Maximize your centrifuge's potential with rotors crafted for efficiency. Tailor your experiments with various capacities and speeds, ensuring optimal sample processing.
Supplier: Eppendorf™ 022654403
Description
- Precision-engineered for optimal sample separation.
- All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121°C, 20 min.).
- Accessories (adaptors and buckets) further customize your centrifuge setup.
- Aerosol-tight and PTFE coated options available.
Certifications
Aerosol-tight rotors are tested and certified by the Health Protection Agency (Porton Down, UK)
Specifications
A-2-MTP - includes wind shield, x2 buckets, and rotor lid | |
2 MTP, PCR, and DWP (max. 0.5 mL) | |
4,680 rpm | |
Centrifuge 5430 / 5430 R | |
Wind Shield, Buckets (x2), and Rotor Lid |
Rotor | |
2,204 x g | |
7.3 cm | |
Fixed |