missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Eppendorf™ 1.5 mL Flex-Tube™ - Microtube

Flex-Tube™ (1.5 mL) are designed for seamless one-handed operation with an easy-to-open, securely closing tube design. Ideal for sample preparation, centrifugation, and storage, these tubes will effortlessly integrate into your workflows for efficient and reliable performance.

Supplier:  Eppendorf™ 022364120

 View more versions of this product

Catalog No. 05-402-96


Explore available promotions

Description

Description

  • Easy-to-use ergonomic design featuring a user-friendly tube lid.
  • Durable polypropylene construction capable of withstanding chemicals, mechanical stress, and extreme temperatures.
  • g-Safe™ technology ensures stability up to 30,000 x g, preventing sample loss during centrifugation and enhancing safety with hazardous samples.
  • Free from slip agents, plasticizers, and biocides, eliminating substances that may compromise bio-assay results.
  • Clear graduation marks allow for visual volume verification during pipetting.
  • Convenient frosted lid and side surface facilitate easy labeling.
  • Functional across a wide temperature range (-86 °C to 100 °C) for versatile applications.
  • Autoclavable up to 121 °C for 20 minutes, ensuring effective sterilization.
  • Available in both Eppendorf Quality and PCR Clean purity grades with lot certification.
Specifications

Specifications

Autoclavable
Clear
Polypropylene
500 Pk.
-122.8°F to +212°F
Human DNA, DNase, RNase, PCR inhibitor-free
PCR Clean (Non-sterile)
0.05 oz.
Cylindrical
Conical
Microtube
1.5 mL
38 mm
Ideal for DNA sample prep prior to PCR
500 Tubes (1 Bag x 500 Tubes)
-86°C to +100°C
Non-sterile
Yes
Snap Cap
1.5 in.
30,000 x g
0.5 mL
Videos
SDS
Documents

Documents

Promotions

Promotions