Eppendorf™ Pipette Pick-a-Pack Sets
4-Pack sets available with Eppendorf Research™ plus single-channel pipettes in a variety of volume ranges.
Supplier:  Eppendorf™ 2231300002
 View more versions of this product
Catalog No. 05-403-151
Explore available promotions
Description
Description
Research Plus Features:
  • Includes four pipetters and one full rack of Eppendorf pipet tips for each pipetter volume
Specifications
Specifications
Autoclavable
Research Plus
2.5, 10, 100, 1000 μL
1
Pipette Package
Pipet 4-pack (0.1–2.5μL, 0.5-10μL, 10–100μL, 100–1,000μL variable-volume pipettes) and epT.I.P.S. pipette tips.