Eppendorf™ Repeater™ M4 Pipets - Trade-In Offer
Exchange any brand pipet with brand new Repeater™ M4 Pipet.
Supplier:  Eppendorf™ 498200421
Catalog No. 05-412-562
Explore available promotions
Description
Description
  • Uses positive displacement principal to precisely and accurately dispense virtually any liquid
  • Has unique volume range of 1μLto 10mL
  • Dispenses aspirated liquid up to 100 times without refilling the tip
  • Step counter supports error-free working
  • Fully emptied Combitip can be easily ejected with one hand using operation lever without directly touching the tip
For repetitive liquid handling
Specifications
Specifications
Repeater™ M4 Pipets Trade-In Offer
Repetitive Liquid Handling
Repeating Pipette
1 to 10 mL
Videos
SDS
Documents
Documents
Accessories