Eppendorf™ Research plus™ Adjustable Volume Multichannel Pipetters Trade-In
Volume adjustment feature allows desired volume to be reached in a few quick turns
Supplier:  Eppendorf™ 2231300045
 View more versions of this product
Catalog No. TI13690048
Explore available promotions
Description
Description
  • Volume adjustment feature allows desired volume to be reached in a few quick turns
  • Control button and ejector's low operating force are positioned for ergonomics
  • 4-digit, magnifying-shape volume display
  • Calibration display and adjustment opening for specific liquids and volumes
  • Ultra light PerfectPiston™ piston system constructed of chemically resistant Fortron™ (not available in 2.5μL, 10μL, 5mL, and 10mL pipetters)
  • Quick connection clip allows removal of lower part in two quick steps (not available in 5 and 10mL pipetters)
  • Spring loaded tip cone promotes tight fit to the tip (not available in 5 and 10mL pipetters)
  • Color indicates pipetter's volume
U.S. Patent 7,434,484.
Specifications
Specifications
Yellow
Research™ Plus™
10 to 100 μL
Yellow
8
Adjustable Volume Pipette
10, 50, 100μL