Eppendorf™ Research™ plus, Mechanical Single-Channel Pipettes with Fixed Volume
Resistant to chemical, thermal, and mechanical stress
Supplier:  Eppendorf™ 3124000075
 View more versions of this product
Catalog No. 13-690-041
Explore available promotions
Description
Description
  • Control button and ejector's low operating force are positioned for ergonomics
  • 4-digit, magnifying-shape volume display Calibration display and adjustment opening for specific liquids and volumes Ultra light PerfectPiston™ piston system constructed of chemically resistant Fortron™ Quick connection clip allows removal of lower part in two quick steps (not in 10μL pipetters) Spring loaded tip cone promotes tight fit to the tip Color indicates pipetter's volume
Specifications
Specifications
±0.6%
Yellow
Fixed Volume Pipette
200μL Pipette tip
Autoclavable
200 μL Pipette Tip
100 μL
Spring loaded tip cone, ejector, yellow operating button
Videos
SDS
Documents
Documents
Accessories