Learn More
Eppendorf™ Research™ plus, Mechanical Single-Channel Pipettes with Fixed Volume
Resistant to chemical, thermal, and mechanical stress
Supplier: Eppendorf™ 3124000113
Description
- Control button and ejector's low operating force are positioned for ergonomics
- 4-digit, magnifying-shape volume display Calibration display and adjustment opening for specific liquids and volumes Ultra light PerfectPiston™ piston system constructed of chemically resistant Fortron™ Quick connection clip allows removal of lower part in two quick steps (not in 10μL pipetters) Spring loaded tip cone promotes tight fit to the tip Color indicates pipetter's volume
Spécifications
| ±0.6% | |
| Blue | |
| Fixed Volume Pipette | |
| 1000μL Pipette tip |
| Autoclavable | |
| 1000 μL Pipette Tip | |
| 500 μL | |
| Spring loaded tip cone, ejector, blue operating button |