Learn More
Eppendorf™ Rotor for Centrifuge 5427 R
Eppendorf Rotor for Centrifuge 5427 R redefine efficiency. Tailor your centrifugation with various capacities and speeds. Explore accessory options for enhanced versatility, ensuring precise sample processing for diverse applications.
Supplier: Eppendorf™ 5409702009
Description
- Precision-engineered for optimal sample separation.
- All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121°C, 20 min.).
- Accessories (adaptors and buckets) further customize your centrifuge setup.
- Aerosol-tight options available.
Specifications
| Aerosol-tight Rotor | |
| 25,001 x g | |
| Centrifuge 5427 R | |
| FA-45-24-11 |
| 24 x 1.5 to 2.0 mL Tubes | |
| 16,220 rpm | |
| Aerosol-tight with Rotor Lid |