Learn More
Eppendorf™ Rotor for Centrifuge 5427 R
Eppendorf Rotor for Centrifuge 5427 R redefine efficiency. Tailor your centrifugation with various capacities and speeds. Explore accessory options for enhanced versatility, ensuring precise sample processing for diverse applications.
Supplier:  Eppendorf™ 5409710001
 View more versions of this product
Catalog No. 05-100-159
Only null left
Explore available promotions
Description
Description
  • Precision-engineered for optimal sample separation.
  • All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121°C, 20 min.).
  • Accessories (adaptors and buckets) further customize your centrifuge setup.
  • Aerosol-tight options available.
Specifications
Specifications
Aerosol-tight Rotor
18,213 x g
Centrifuge 5427 R
FA-45-48-11
48 x 1.5 to 2.0 mL Tubes
12,700 rpm
Aerosol-tight with Rotor Lid
Product Suggestions
Product Suggestions