missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ Rotor for Centrifuge 5427 R
Eppendorf Rotor for Centrifuge 5427 R redefine efficiency. Tailor your centrifugation with various capacities and speeds. Explore accessory options for enhanced versatility, ensuring precise sample processing for diverse applications.
Supplier: Eppendorf™ 5409714007
Description
- Precision-engineered for optimal sample separation.
- All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121°C, 20 min.).
- Accessories (adaptors and buckets) further customize your centrifuge setup.
- Aerosol-tight options available.
Specifications
| Rotor | |
| 11,710 x g | |
| Centrifuge 5427 R | |
| 48 |
| 48 x 0.2 mL PCR Tubes | |
| 10,500 rpm | |
| Plastic Rotor Lid | |
| F-45-48-5-PCR |