missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Eppendorf™ Safe-Lock Tubes 1.5 mL - Microtube
Eppendorf™ Safe-Lock Tubes 1.5 mL - Microtube ensures secure sample storage. With a patented Safe-Lock design, these microtubes prevent accidental opening, safeguarding samples. Ideal for diverse applications, they offer reliability, durability, and ease of use in a 1.5 mL capacity.
Supplier: Eppendorf™ 022600028
Description
- The hinged Eppendorf Safe-Lock lid prevents inadvertent lid opening during incubation and storage.
- g-Safe™ technology, depending on the Safe-Lock Tube variant (refer to the manual), ensures outstanding centrifugation stability up to 30,000 x g, preventing sample loss due to tube breakage and providing additional safety when handling hazardous samples.
- Manufactured without the use of slip agents, plasticizers, and biocides – substances demonstrated to leach from plastic consumables into the sample and adversely impact bio-assay results.
- Polypropylene construction offers robust resistance to chemicals, mechanical stress, and extreme temperatures.
- Precision lid sealing minimizes evaporation rates during extended storage periods.
- The generously sized frosted lid and side surface facilitate easy labeling.
- Assured functionality in a temperature range from -86 °C to 100 °C.
- Autoclavable when open (121 °C, 20 min) for reliable sterilization.
- Available in lot-certified PCR clean, Eppendorf Forensic DNA Grade, and Biopur™ quality for tested purity.
Order Info
Supplied individually wrapped in your choice of three sizes
Specifications
10 mm | |
Quality assurance card | |
Autoclavable | |
Clear | |
Polypropylene | |
100 Tubes, Individually Wrapped | |
-86°C to +100°C | |
Sterile | |
Yes | |
Safe-lock Lid | |
1.5 in. |
22,000 x g | |
Polypropylene | |
1.5 mL | |
38 mm | |
100 Pk. | |
-122.8°F to +212°F | |
Free of plasticizers, biocides or mold release agents | |
Biopur™ (Sterile) | |
0.05 oz. | |
Cylindrical | |
Conical |