MilliporeSigma™ Supelco™ Eppendorf™ TC-45 Temperature Controller
This low-cost controller will control the CH-30 Eppendorf heater from 30-65°C. Temperature is set via DIP switches in 5°C increments.
Supplier:  MilliporeSigma™ Supelco™ 500763
Catalog No. 111007024
Explore available promotions
Description
Description
Specifications
Specifications
HPLC
AC/DC input 240 V AC
Videos
SDS
Documents
Documents
Promotions
Promotions
Eppendorf is a registered trademark of Eppendorf AG