Eppendorf™ twin.tec™ PCR Plates 384-well, 45 μL - PCR Plates
Eppendorf's twin.tec™ PCR Plates feature a polycarbonate frame and thin-walled polypropylene wells ensuring optimal heat transfer and high rigidity. With raised well rims, these plates enable an effective seal, minimizing the risk of cross-contamination and contributing to consistently reliable results.
Supplier:  Eppendorf™ 951020745
 View more versions of this product
Catalog No. E951020745
Explore available promotions
Description
Description
  • One-piece design: combining a polycarbonate frame and polypropylene wells for optimum performance.
  • Extremely thin-walled polypropylene wells guarantee optimum heat transfer to the sample.
  • Exceptionally solid and torque-resistant polycarbonate frame.
  • Improved well-to-well tolerance.
  • Cut-away corner and alphanumeric grid-referencing.
  • Raised well rims for effective sealing, also reduces risk of cross-contamination.
  • Batch-tested and certified free of DNA, DNase, RNase and PCR inhibitors (PCR clean).
  • Safe Code option provides 3-level coding for safe sample and vessel identification.
  • ECC 200 error correction function enables safe code reading with up to 30% damaged codes.
Certifications
Certified free of human DNA, DNase, RNase, and PCR inhibitors
Specifications
Specifications
No Barcode
Standard
Autoclavable
Red
Automated Systems
45 μL
Non-sterile
2250 x g
25 Pk.
Colorless
Free of Human DNA, DNase, RNase and PCR Inhibitors
16 x 24
25 Plates (5 Bags x 5 Plates)
PCR Plate
Polypropylene, Polycarbonate
384
Round Bottom (U-well)
PCR Clean (Non-sterile)
Yes