missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Eppendorf™ epT.I.P.S.™ 384 Reloads - Micropipette Tips

Eppendorf epT.I.P.S.™ 384 Reloads provide high-precision micropipette tips in 'Eppendorf Quality' and 'PCR clean' grades, tailored for 384-well plate applications, ensuring accuracy, reduced waste, and compatibility in high-throughput labs.

$790.16 - $975.60

Specifications

Filtered Non-filtered
Sterility Non-sterile
Color Colorless
Autoclavable Autoclavable
Model epT.I.P.S.™ 384 Reloads
View More Specs

Products 4
Catalog Number Mfr. No. Clean Claims Volume (Metric) Color Code Length (Metric) Price Quantity  
Catalog Number Mfr. No. Clean Claims Volume (Metric) Color Code Length (Metric) Price Quantity  
05413607
View Documents
Eppendorf™
0030076001
PCR Clean (Non-sterile) 0.1 to 20 μL Light Pink 42 mm
Case of 3840 Each for $975.60
 
05413610
View Documents
Eppendorf™
0030076010
PCR Clean (Non-sterile) 5 to 100 μL Light Yellow 53 mm
Case of 3840 Each for $864.95
 
05413608
View Documents
Eppendorf™
0030076044
Eppendorf Quality (Non-sterile) 0.1 to 20 μL Light Pink 42 mm
Case of 3840 Each for $878.98
 
05413611
View Documents
Eppendorf™
0030076052
Eppendorf Quality (Non-sterile) 5 to 100 μL Light Yellow 53 mm
Case of 3840 Each for $790.16
 
Description

Description

  • epT.I.P.S. 384 pipette tips build a system together with the Research plus and Xplorer plus 16- and 24-channel pipettes as well as 20 μL and 100 μL Move It adjustable tip spacing pipettes for utmost safety and reliability
  • Designed for a cone distance of 4.5 mm
  • Innovative SOFTattach technology allows for reliable tip fit and tightness
  • Fill a whole 384-well plate without loosening pipette tips
  • Perfect tip alignment by extraordinary coaxiality for safe plate maneuvering and liquid transfer
  • The tip shape of the epT.I.P.S. 384 is adjusted to perfectly fit the wells of 384-well plates
  • Tip attachment forces reduced by 40% per cone compared to 8- and 12-channel pipettes
  • Tip ejection forces in 16- and 24-channel pipettes reduced by SOFTeject for ergonomic working conditions
  • Available as reload, and set
  • Available in purity grades of Eppendorf Quality and PCR clean
  • ACT™-labeled by non-profit My Green Lab™ to help you make an informed product choice when it comes to sustainability
Specifications

Specifications

Non-filtered
Colorless
epT.I.P.S.™ 384 Reloads
Universal
Universal
3840 Tips
Single-use
epT.I.P.S.
Non-sterile
Autoclavable
Room temperature
Refill
3,840 Tips (10 Trays x 384 Tips)
Micropipette Tips
Air-cushion System
Polypropylene
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

The pipette tips of the :Productname: product group with the respective product numbers are designed and constructed for low-contamination transfer of liquids, in particular for samples from the human body and for reagents within the scope of an in-vitro diagnostic application in order to allow the in-vitro diagnostic medical device to be used as intended. The above mentioned :Productname: are in-vitro diagnostic devices according to Directive 98/79/EC of the European Parliament and the Council dated October 27, 1998. They are intended exclusively for single use and for application by qualified staff.