Eppendorf™ epT.I.P.S.™ Racks - Pipette Tips
Eppendorf epT.I.P.S.™ Racks feature high-precision pipette tips in 'Eppendorf Quality', 'PCR clean', 'Biopur™', and 'Sterile' purity grades, in disposable racks designed for optimal organization and user convenience in demanding laboratory environments.
Supplier:  Eppendorf™ 0030075234
 View more versions of this product
Catalog No. 05413931
Explore available promotions
Description
Description
  • Long, narrow tips reach the base of narrow and high vessels without touching the top of the vessel edge
  • Modern slim rack design with reclosable lid
  • Improved sustainability through 35% lower polypropylene consumption per rack
  • ACT™-labeled by non-profit My Green Lab™ to help you make an informed product choice when it comes to sustainability
Specifications
Specifications
Replaces Order No. 0030075030
Non-filtered
Colorless
53 mm
Biopur™ (Sterile)
Eppendorf Pipettes, Non-Eppendorf Pipettes
Narrow
480 Tips (5 x 96)
Racks, Lid
Single-use
Yes
Polypropylene
2 to 200 μL
Sterile
2.08 in.
ISO 9001, ISO 13485, ISO 14001, ISO 14644-1, EN ISO 8655, Certificate of Quality, Lot Specific Certificates Available
Cell/Molecular Biology, Medical, Pharmaceutical, and Food Industries
Rack
480 Tips (5 Racks x 96 Tips)
Pipette Tips
Yellow
Air-cushion System
epT.I.P.S.