Eppendorf™ epT.I.P.S.™ Racks - Pipette Tips Edge
Edge
Eppendorf epT.I.P.S.™ Racks feature high-precision pipette tips in 'Eppendorf Quality', 'PCR clean', 'Biopur™', and 'Sterile' purity grades, in disposable racks designed for optimal organization and user convenience in demanding laboratory environments.
Supplier:  Eppendorf™ 0030071654
 View more versions of this product
Catalog No. 05414001
Edge
Explore available promotions
Description
Description
  • Long, narrow tips reach the base of narrow and high vessels without touching the top of the vessel edge
  • Modern slim rack design with reclosable lid
  • Improved sustainability through 35% lower polypropylene consumption per rack
  • ACT™-labeled by non-profit My Green Lab™ to help you make an informed product choice when it comes to sustainability
Specifications
Specifications
0.5 to 10 mL
Non-sterile
Turquoise
165 mm
Cell/Molecular Biology, Medical, Pharmaceutical, and Food Industries
Rack
120 Tips (5 Racks x 24 Tips)
Turquoise
Air-cushion System
epT.I.P.S.
Non-filtered
Turquoise
6.5 in.
Eppendorf Quality (Non-sterile)
Eppendorf Pipettes, Non-Eppendorf Pipettes
Narrow
Pipette Tips
Single-use
Yes
Polypropylene