Supertek™ Equal Mass Set DFS Item
To demonstrate the inverse relationship between density and length.
Supplier:  Supertek™ PH10045
Catalog No. S012344
Explore available promotions
Description
Description
  • Demonstrate the inverse relationship between density and length.
  • The specimens include: Aluminum, Copper, Nylon, Polyvinylchloride and Polyethylene.
  • All are 12 mm in diameter with 15 g mass
  • Length range from 15 to 100 mm.
  • Instructions are included.
Specifications
Specifications
Equal Mass Cylinders Set